Gene Transcripts

Gene Symbol
Gripap1
Entrez Gene ID
116493
Gene Name
GRIP1 associated protein 1
Gene Aliases
Grasp1
Chromosome Location
Chr. X: 15523929 - 15553702 on Build Rnor_6.0
UniGene ID
Rn.53868
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053807.1 NP_446259.1 3 202
GenBank mRNA AF274057.1 - 3 202
AF274058.1 - 3 202

View Full Product Details