Gene Transcripts

Gene Symbol
Pip4k2a
Entrez Gene ID
116723
Gene Name
phosphatidylinositol-5-phosphate 4-kinase, type II, alpha
Gene Aliases
Pip5k2a
Chromosome Location
Chr. 17: 85533056 - 85748843 on Build Rnor_6.0
UniGene ID
Rn.24177
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053926.2 NP_446378.1 6 806
GenBank mRNA AB032899.1 - 6 806

View Full Product Details