Gene Transcripts

Gene Symbol
Acot8
Entrez Gene ID
170588
Gene Name
acyl-CoA thioesterase 8
Gene Aliases
Pte1
Chromosome Location
Chr. 3: 161260837 - 161272495 on Build Rnor_6.0
UniGene ID
Rn.161868
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130756.3 NP_570112.3 2 534
GenBank mRNA AF452100.1 - 2 184
BC078751.1 - 2 528 7114090
CV115391.1 - 2 534

View Full Product Details