Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014553.2 | NP_055368.1 | 6, 7 | 749 | |
XM_017003902.1 | XP_016859391.1 | 6, 7 | 757 | ||
XM_017003904.1 | XP_016859393.1 | 6, 7 | 757 | ||
XM_017003905.1 | XP_016859394.1 | 6, 7 | 757 | ||
GenBank mRNA | AF198488.1 | - | 6, 7 | 746 | |
AL137740.1 | - | 6, 7 | 749 | ||
BC064698.1 | - | 6, 7 | 749 | 30347863 |