Gene Transcripts
- Gene Symbol
- Prol1
- Entrez Gene ID
- 65182
- Gene Name
- proline rich, lacrimal 1
- Gene Aliases
- Muc10, RSM-1
- Chromosome Location
- Chr. 14: 21338464 - 21353533 on Build Rnor_6.0
- UniGene ID
- Rn.10113
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133513.1 |
NP_598197.1
|
3 |
229 |
|
GenBank mRNA |
U03407.1 |
- |
3 |
229 |
|
View Full Product Details