Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133516.2 | NP_598200.1 | 2 | 279 | |
GenBank mRNA | AF159097.1 | - | 1 | 48 | |
X01115.1 | - | 2 | 279 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008773873.1 | XP_008772095.1 | 2 | 314 |