Gene Transcripts

Gene Symbol
Mrpl17
Entrez Gene ID
171061
Gene Name
mitochondrial ribosomal protein L17
Gene Aliases
Chromosome Location
Chr. 1: 170652295 - 170653869 on Build Rnor_6.0
UniGene ID
Rn.40354
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133539.1 NP_598223.1 3 238
GenBank mRNA BC058447.1 - 3 283
FQ212468.1 - 3 344
FQ229319.1 - 3 296
U53512.1 - 3 238

View Full Product Details