Gene Transcripts

Gene Symbol
Calcb
Entrez Gene ID
171519
Gene Name
calcitonin-related polypeptide, beta
Gene Aliases
Chromosome Location
Chr. 1: 184271033 - 184276480 on Build Rnor_6.0
UniGene ID
Rn.88080
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138513.1 NP_612522.1 2 181
XM_006230075.2 XP_006230137.1 4 655
XM_008759674.1 XP_008757896.1 5 510
GenBank mRNA M11596.1 - 2 181

View Full Product Details