Gene Transcripts

Gene Symbol
Zp4
Entrez Gene ID
282833
Gene Name
zona pellucida glycoprotein 4
Gene Aliases
Zp-4
Chromosome Location
Chr. 17: 65497359 - 65503912 on Build Rnor_6.0
UniGene ID
Rn.91141
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172330.3 NP_758833.2 2 245
GenBank mRNA AF456325.1 - 2 246

View Full Product Details