Gene Transcripts

Gene Symbol
Spats1
Entrez Gene ID
301255
Gene Name
spermatogenesis associated, serine-rich 1
Gene Aliases
Srsp1
Chromosome Location
Chr. 9: 17911950 - 17947101 on Build Rnor_6.0
UniGene ID
Rn.40672
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181376.2 NP_852041.1 3 377
GenBank mRNA AY157978.1 - 3 377

View Full Product Details