Gene Transcripts

Gene Symbol
Retn
Entrez Gene ID
246250
Gene Name
resistin
Gene Aliases
Chromosome Location
Chr. 12: 2201909 - 2203649 on Build Rnor_6.0
UniGene ID
Rn.16746
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144741.1 NP_653342.1 2 153
GenBank mRNA AF378366.1 - 2 153

View Full Product Details