Gene Transcripts

Gene Symbol
Cbx2
Entrez Gene ID
12416
Gene Name
chromobox 2
Gene Aliases
M33, MOD2, pc
Chromosome Location
Chr. 11: 119023019 - 119031274 on Build GRCm38
UniGene ID
Mm.14547
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007623.2 NP_031649.2 5 562
GenBank mRNA AK157869.1 - 5 538
BC035199.1 - 5 562 5137505
X62537.1 - 5 629
Y13274.1 - 5 486

View Full Product Details