Gene Transcripts

Gene Symbol
PLXDC1
Entrez Gene ID
57125
Gene Name
plexin domain containing 1
Gene Aliases
TEM3, TEM7
Chromosome Location
Chr. 17: 39063303 - 39151649 on Build GRCh38
UniGene ID
Hs.125036
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020405.4 NP_065138.2 14 5933
GenBank mRNA AF070526.1 - 1 1633
AF378753.1 - 14 5802

View Full Product Details