Gene Transcripts

Gene Symbol
Nedd1
Entrez Gene ID
17997
Gene Name
neural precursor cell expressed, developmentally down-regulated gene 1
Gene Aliases
Chromosome Location
Chr. 10: 92684745 - 92723682 on Build GRCm38
UniGene ID
Mm.2998
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008682.2 NP_032708.2 15 3110
GenBank mRNA AK010591.1 - 15 3098
AK048393.1 - 15 3062
AK153746.1 - 15 3112
BC052430.1 - 15 3079 6836583
BC066870.1 - 15 3047 30476383
D10712.1 - 15 3052

View Full Product Details