Gene Transcripts

Gene Symbol
Rab23
Entrez Gene ID
19335
Gene Name
RAB23, member RAS oncogene family
Gene Aliases
AW545388, opb, opb2
Chromosome Location
Chr. 1: 33719896 - 33742564 on Build GRCm38
UniGene ID
Mm.86744
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159729.1 NP_001153201.1 8 1801
NM_008999.4 NP_033025.2 8 1913
GenBank mRNA AK016493.1 - 8 1813
AK047068.1 - 8 1914
AK082017.1 - 8 1877
AK164057.1 - 8 1998

View Full Product Details