Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009077.2 | NP_033103.2 | 6 | 627 | |
GenBank mRNA | AK007263.1 | - | 6 | 597 | |
AK009044.1 | - | 6 | 598 | ||
AK010510.1 | - | 6 | 595 | ||
AK012580.1 | - | 6 | 595 | ||
AK145649.1 | - | 6 | 628 | ||
AK151667.1 | - | 6 | 597 | ||
AK153172.1 | - | 6 | 595 | ||
AK166803.1 | - | 6 | 597 | ||
AK168288.1 | - | 6 | 595 | ||
AK169659.1 | - | 6 | 595 | ||
BC072558.1 | - | 2936 | |||
BC081468.1 | - | 6 | 585 | 6823249 | |
BC082290.1 | - | 6 | 581 | 6485569 | |
BC091732.1 | - | 5 | 573 | 2655484 | |
L04128.1 | - | 5 | 587 |