Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009294.3 | NP_033320.1 | 2, 3 | 371 | |
GenBank mRNA | AK145909.1 | - | 2, 3 | 390 | |
AK161971.1 | - | 2, 3 | 364 | ||
BC005791.1 | - | 2, 3 | 211 | 3603665 | |
BC011491.1 | - | 2, 3 | 371 | 4016871 | |
U76832.1 | - | 2, 3 | 140 |