Gene Transcripts

Gene Symbol
Supt6
Entrez Gene ID
20926
Gene Name
suppressor of Ty 6
Gene Aliases
5131400N11Rik, AI132449, SPT6, Supt6h, mKIAA0162
Chromosome Location
Chr. 11: 78206749 - 78245703 on Build GRCm38
UniGene ID
Mm.20755
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009297.2 NP_033323.2 3 311
GenBank mRNA AK136094.1 - 951
AK170533.1 - 3 310
BC072657.1 - 3 231 30536338
U40375.1 - 3 309

View Full Product Details