Gene Transcripts

Gene Symbol
Tacr2
Entrez Gene ID
21337
Gene Name
tachykinin receptor 2
Gene Aliases
Nk2r, Skr, Tac2r
Chromosome Location
Chr. 10: 62252438 - 62265990 on Build GRCm38
UniGene ID
Mm.8054
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009314.4 NP_033340.3 1, 2 780
GenBank mRNA AK033443.1 - 1, 2 463
BC106959.1 - 1, 2 429 40039032
X62933.1 - 1, 2 780

View Full Product Details