Gene Transcripts
- Gene Symbol
- Ssr4
- Entrez Gene ID
- 20832
- Gene Name
- signal sequence receptor, delta
- Gene Aliases
- AW743063, C78062, Trap
- Chromosome Location
- Chr. X: 73787028 - 73790828 on Build GRCm38
- UniGene ID
- Mm.831
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001166480.1 |
NP_001159952.1
|
4, 5 |
567 |
|
NM_009279.3 |
NP_033305.1
|
4, 5 |
449 |
|
GenBank mRNA |
AK003044.1 |
- |
4, 5 |
429 |
|
AK007921.1 |
- |
4, 5 |
405 |
|
AK013723.1 |
- |
4, 5 |
411 |
|
AK160089.1 |
- |
4, 5 |
415 |
|
AK191259.1 |
- |
2, 3 |
149 |
|
BC008172.1 |
- |
4, 5 |
370 |
3481948 |
BC019419.1 |
- |
3, 4 |
1900 |
|
BI989805.1 |
- |
4, 5 |
567 |
|
BQ747160.1 |
- |
4, 5 |
352 |
|
X90582.1 |
- |
4, 5 |
530 |
|
View Full Product Details