Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009355.2 | NP_033381.1 | 2 | 281 | |
GenBank mRNA | AB008910.1 | - | 2 | 281 | |
BC049616.1 | - | 2 | 276 | 6773970 | |
BC115665.2 | - | 2 | 280 | 40103370 | |
BC115666.2 | - | 2 | 280 | 40103373 |