Gene Transcripts

Gene Symbol
Tnfsf8
Entrez Gene ID
21949
Gene Name
tumor necrosis factor (ligand) superfamily, member 8
Gene Aliases
CD153, CD30LG, Cd30l
Chromosome Location
Chr. 4: 63832820 - 63861681 on Build GRCm38
UniGene ID
Mm.4664
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009403.3 NP_033429.1 4 1848
GenBank mRNA AK030916.1 - 4 1849
AK157270.1 - 4 1601
L09754.1 - 4 1737

View Full Product Details