Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159627.1 | NP_001153099.1 | 3 | 606 | |
NM_001159628.1 | NP_001153100.1 | 3 | 614 | ||
NM_010417.2 | NP_034547.2 | 3 | 614 | ||
NM_181273.4 | NP_851790.1 | 3 | 606 | ||
GenBank mRNA | AF082567.1 | - | 3 | 726 | |
AK032895.1 | - | 3 | 526 | ||
AK081330.1 | - | 3 | 582 | ||
AK086923.1 | - | 3 | 525 | ||
AK129194.1 | - | 3 | 562 | ||
BC048237.1 | - | 3 | 454 | 6474714 | |
BC054442.1 | - | 3 | 454 | 6474713 |