Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011288.1 | NP_035418.1 | 3 | 218 | |
GenBank mRNA | AB049646.1 | - | 3 | 172 | |
AK086805.1 | - | 3 | 172 | ||
BC038626.1 | - | 3 | 190 | 4008503 | |
BC081460.1 | - | 3 | 158 | 6814073 | |
U84902.1 | - | 3 | 160 |