Gene Transcripts
- Gene Symbol
- Saa2
- Entrez Gene ID
- 20209
- Gene Name
- serum amyloid A 2
- Gene Aliases
- AW111173, Saa-2, Saa1
- Chromosome Location
- Chr. 7: 46751777 - 46754314 on Build GRCm38
- UniGene ID
- Mm.200941
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M11130.1 |
- |
4 |
540 |
|
View Full Product Details