Gene Transcripts
- Gene Symbol
- Zfp59
- Entrez Gene ID
- 22717
- Gene Name
- zinc finger protein 59
- Gene Aliases
- Mfg-2, Mfg2
- Chromosome Location
- Chr. 7: 27838540 - 27856793 on Build GRCm38
- UniGene ID
- Mm.379046
- Species
- Mus musculus
- Gene Symbol
- Gm7452
- Entrez Gene ID
- 665024
- Gene Name
- predicted pseudogene 7452
- Gene Aliases
- EG665024
- Chromosome Location
- Chr. 6: 36956654 - 36960430 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC127053.1 |
- |
1 |
294 |
|
View Full Product Details