Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011775.6 | NP_035905.1 | 5, 6 | 490 | |
GenBank mRNA | AK135903.1 | - | 5, 6 | 490 | |
AK138699.1 | - | 5, 6 | 491 | ||
BC071183.1 | - | 5, 6 | 472 | 30473497 | |
M34148.1 | - | 5, 6 | 490 |