Gene Transcripts

Gene Symbol
Rgs7
Entrez Gene ID
24012
Gene Name
regulator of G protein signaling 7
Gene Aliases
Chromosome Location
Chr. 1: 175059076 - 175493238 on Build GRCm38
UniGene ID
Mm.7956
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199003.1 NP_001185932.1 7 759
NM_011880.3 NP_036010.2 7 759
GenBank mRNA AF011360.1 - 7 604
BC051133.1 - 7 714 6588901

View Full Product Details