Gene Transcripts
- Gene Symbol
- Nrk
- Entrez Gene ID
- 27206
- Gene Name
- Nik related kinase
- Gene Aliases
- AA407323, AA409510, AI225895, AV168563, AW538185, Nesk
- Chromosome Location
- Chr. X: 138914312 - 139010532 on Build GRCm38
- UniGene ID
- Mm.482319
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB020741.1 |
- |
28 |
4883 |
|
View Full Product Details