Gene Transcripts

Gene Symbol
Stra13
Entrez Gene ID
20892
Gene Name
stimulated by retinoic acid 13
Gene Aliases
Chromosome Location
Chr. 11: 120710941 - 120713903 on Build GRCm38
UniGene ID
Mm.259716
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016665.2 NP_057874.2 3, 4 175
GenBank mRNA AK076181.1 - 2, 3 234
AK080500.1 - 3, 4 147
AK170126.1 - 3, 4 147
BC052335.1 - 4, 5 425
CA320983.1 - 2, 3 114
U95003.1 - 4, 5 239
U95005.1 - 4, 5 413

View Full Product Details