Gene Transcripts

Gene Symbol
Dclre1a
Entrez Gene ID
55947
Gene Name
DNA cross-link repair 1A, PSO2 homolog (S. cerevisiae)
Gene Aliases
2810043H12Rik, AU022226, Pso2, Smn1a, Snm1, mKIAA0086
Chromosome Location
Chr. 19: 56529161 - 56548284 on Build GRCm38
UniGene ID
Mm.2805
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF241240.1 - 1 286

View Full Product Details