Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019734.2 | NP_062708.1 | 4 | 339 | |
GenBank mRNA | AF157500.1 | - | 4 | 269 | |
AF352179.1 | - | 4 | 281 | ||
AK075666.1 | - | 4 | 254 | ||
AK075919.1 | - | 4 | 293 | ||
AK077721.1 | - | 4 | 292 | ||
AK090314.1 | - | 4 | 289 | ||
AK150567.1 | - | 4 | 252 | ||
AK151208.1 | - | 4 | 340 | ||
AK151312.1 | - | 4 | 252 | ||
AK151478.1 | - | 4 | 330 | ||
AK152305.1 | - | 4 | 291 | ||
AK152811.1 | - | 4 | 252 | ||
AK152920.1 | - | 4 | 299 | ||
AK153101.1 | - | 4 | 299 | ||
AK153297.1 | - | 4 | 253 | ||
AK159556.1 | - | 4 | 261 | ||
AK166757.1 | - | 4 | 271 | ||
BC003204.1 | - | 4 | 259 | 3587042 | |
BY160973.1 | - | 4 | 340 |