Gene Transcripts
- Gene Symbol
- Xpo4
- Entrez Gene ID
- 57258
- Gene Name
- exportin 4
- Gene Aliases
- B430309A01Rik, mKIAA1721
- Chromosome Location
- Chr. 14: 57577520 - 57664985 on Build GRCm38
- UniGene ID
- Mm.202747
- Species
- Mus musculus
- Gene Symbol
- Lats2
- Entrez Gene ID
- 50523
- Gene Name
- large tumor suppressor 2
- Gene Aliases
- 4932411G09Rik, AV277261, AW228608
- Chromosome Location
- Chr. 14: 57689662 - 57746123 on Build GRCm38
- UniGene ID
- Mm.347899
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029242.1 |
- |
|
3059 |
|
View Full Product Details