Gene Transcripts
- Gene Symbol
- Aox3
- Entrez Gene ID
- 71724
- Gene Name
- aldehyde oxidase 3
- Gene Aliases
- 1200011D03Rik, AI326299, Aoh1, Aoh2
- Chromosome Location
- Chr. 1: 58113136 - 58200452 on Build GRCm38
- UniGene ID
- Mm.20108
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF172276.2 |
- |
31 |
3663 |
|
AY347569.1 |
- |
31 |
3464 |
|
AY347570.1 |
- |
31 |
3464 |
|
View Full Product Details