Gene Transcripts

Gene Symbol
Lrrk2
Entrez Gene ID
66725
Gene Name
leucine-rich repeat kinase 2
Gene Aliases
4921513O20Rik, 9330188B09Rik, AW561911, D630001M17Rik, Gm927, cI-46
Chromosome Location
Chr. 15: 91672821 - 91816124 on Build GRCm38
UniGene ID
Mm.37558
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025730.3 NP_080006.3 43, 44 6446
GenBank mRNA AK014886.1 - 4, 5 505
AK014938.1 - 4, 5 518
AY792512.1 - 43, 44 6389
BC034074.1 - 9, 10 1329

View Full Product Details