Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025770.3 | NP_080046.3 | 3, 4 | 269 | |
GenBank mRNA | AB079383.1 | - | 2, 3 | 225 | |
AB091691.1 | - | 2, 3 | 246 | ||
AK009076.1 | - | 3, 4 | 253 | ||
AK017355.1 | - | 3, 4 | 251 | ||
AK077333.1 | - | 3, 4 | 242 | ||
BB843097.1 | - | 3, 4 | 269 | ||
BC023455.1 | - | 2, 3 | 245 | 5249268 |