Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163687.1 | NP_001157159.1 | 3 | 472 | |
NM_025972.4 | NP_080248.2 | 3 | 478 | ||
GenBank mRNA | AB162194.1 | - | 3 | 444 | |
AK008776.1 | - | 3 | 480 | ||
AK017141.1 | - | 3 | 457 | ||
AK133485.1 | - | 3 | 463 | ||
AK133658.1 | - | 3 | 480 |