Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026068.2 | NP_080344.2 | 3 | 733 | |
GenBank mRNA | AK005655.1 | - | 3 | 142 | |
AK013990.1 | - | 3 | 174 | ||
AK045566.1 | - | 1 | 1211 | ||
BC019685.1 | - | 3 | 203 | 3983617 | |
BC051929.1 | - | 3 | 733 | 5709831 |