Gene Transcripts
- Gene Symbol
- TYRO3
- Entrez Gene ID
- 7301
- Gene Name
- TYRO3 protein tyrosine kinase
- Gene Aliases
- BYK, Dtk, Etk-2, RSE, Rek, Sky, Tif
- Chromosome Location
- Chr. 15: 41559022 - 41579338 on Build GRCh38
- UniGene ID
- Hs.381282
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006293.3 |
NP_006284.2
|
3, 4 |
653 |
|
XM_017022542.1 |
XP_016878031.1
|
3, 4 |
618 |
|
XM_017022543.1 |
XP_016878032.1
|
3, 4 |
454 |
|
GenBank mRNA |
AB209431.1 |
- |
3, 4 |
682 |
|
BC034960.1 |
- |
3, 4 |
435 |
|
BC049368.1 |
- |
3, 4 |
427 |
4821587 |
BC051756.1 |
- |
3, 4 |
432 |
6095596 |
D17517.1 |
- |
3, 4 |
641 |
|
D50479.1 |
- |
3, 4 |
413 |
|
U05682.1 |
- |
3, 4 |
423 |
|
View Full Product Details