Gene Transcripts

Gene Symbol
Reg4
Entrez Gene ID
67709
Gene Name
regenerating islet-derived family, member 4
Gene Aliases
2010002L15Rik, GISP, RELP
Chromosome Location
Chr. 3: 98222156 - 98236748 on Build GRCm38
UniGene ID
Mm.46306
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026328.2 NP_080604.2 5 531
GenBank mRNA AK008049.1 - 5 533
AK008438.1 - 5 534
AK160329.1 - 5 529
AK162316.1 - 5 532
BC019465.1 - 5 514 4216795

View Full Product Details