Gene Transcripts

Gene Symbol
N6amt2
Entrez Gene ID
68043
Gene Name
N-6 adenine-specific DNA methyltransferase 2 (putative)
Gene Aliases
2510005D08Rik, AW045965, Ayu21-96, Gt(pU21)96Imeg, GtAyu21-96
Chromosome Location
Chr. 14: 57549596 - 57571569 on Build GRCm38
UniGene ID
Mm.45225
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026526.2 NP_080802.1 2 117
GenBank mRNA AK005312.1 - 2 111
AK010911.1 - 1 112
AK012083.1 - 2 118
BC051925.1 - 2 108 5691329

View Full Product Details