Gene Transcripts

Gene Symbol
Uckl1
Entrez Gene ID
68556
Gene Name
uridine-cytidine kinase 1-like 1
Gene Aliases
1110007H10Rik, Urkl1
Chromosome Location
Chr. 2: 181569149 - 181581973 on Build GRCm38
UniGene ID
Mm.232400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026765.3 NP_081041.2 13 1339
GenBank mRNA AK003541.1 - 3 185
BC016535.1 - 13 1339 2655873

View Full Product Details