Gene Transcripts
- Gene Symbol
- Spcs1
- Entrez Gene ID
- 69019
- Gene Name
- signal peptidase complex subunit 1 homolog (S. cerevisiae)
- Gene Aliases
- 1810004F21Rik
- Chromosome Location
- Chr. 14: 30999826 - 31001666 on Build GRCm38
- UniGene ID
- Mm.28852
- Species
- Mus musculus
- Gene Symbol
- Gtpbp8
- Entrez Gene ID
- 66067
- Gene Name
- GTP-binding protein 8 (putative)
- Gene Aliases
- 0610037H22Rik
- Chromosome Location
- Chr. 16: 44738879 - 44746377 on Build GRCm38
- UniGene ID
- Mm.34520
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK084919.1 |
- |
1 |
1213 |
|
View Full Product Details