Gene Transcripts

Gene Symbol
Sectm1b
Entrez Gene ID
58210
Gene Name
secreted and transmembrane 1B
Gene Aliases
1810003C24Rik, K12, Sectm1
Chromosome Location
Chr. 11: 121053423 - 121063569 on Build GRCm38
UniGene ID
Mm.116986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026907.3 NP_081183.3 5 1554
GenBank mRNA AK029082.1 - 5 1555
AK143807.1 - 5 1540

View Full Product Details