Gene Transcripts

Gene Symbol
Lyrm1
Entrez Gene ID
73919
Gene Name
LYR motif containing 1
Gene Aliases
1110065L10Rik, 2310004B22Rik, 4930404J24Rik
Chromosome Location
Chr. 7: 119895807 - 119956203 on Build GRCm38
UniGene ID
Mm.291751
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285959.1 NP_001272888.1 4 347
NM_001285960.1 NP_001272889.1 4 480
NM_029610.3 NP_083886.1 4 731
GenBank mRNA AK009135.1 - 4 734
AK015083.1 - 5 539
AK157931.1 - 349
BC011339.1 - 4 521 3978191
BY752674.1 - 4 349
DV043140.1 - 3 505

View Full Product Details