Gene Transcripts

Gene Symbol
Nus1
Entrez Gene ID
52014
Gene Name
nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae)
Gene Aliases
1600027K07Rik, AU019165, AW538011, BC003223, D10Ertd438e
Chromosome Location
Chr. 10: 52417547 - 52440192 on Build GRCm38
UniGene ID
Mm.199964
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030250.2 NP_084526.1 3 830
GenBank mRNA AK155620.1 - 3 831
AK167741.1 - 3 819
AK214556.1 - 1 31
BC003223.1 - 3 789 3482163
BC018372.1 - 3 805 3499868

View Full Product Details