Gene Transcripts
- Gene Symbol
- Rtn3
- Entrez Gene ID
- 20168
- Gene Name
- reticulon 3
- Gene Aliases
- RTN3-A1
- Chromosome Location
- Chr. 19: 7425895 - 7483291 on Build GRCm38
- UniGene ID
- Mm.246990
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003933.2 |
NP_001003933.1
|
3, 4 |
2680 |
|
NM_001003934.2 |
NP_001003934.1
|
4, 5 |
2737 |
|
NM_001271486.1 |
NP_001258415.1
|
4, 5 |
1774 |
|
NM_001271487.1 |
NP_001258416.1
|
3, 4 |
613 |
|
NM_053076.3 |
NP_444306.1
|
2, 3 |
556 |
|
GenBank mRNA |
AB046114.1 |
- |
2, 3 |
550 |
|
AF195940.1 |
- |
2, 3 |
500 |
|
AK032109.1 |
- |
4, 5 |
1751 |
|
AK036892.1 |
- |
2, 3 |
522 |
|
AK049845.1 |
- |
2, 3 |
488 |
|
AK075883.1 |
- |
2, 3 |
485 |
|
AK088670.1 |
- |
2, 3 |
488 |
|
AK146505.1 |
- |
2, 3 |
364 |
|
AK148792.1 |
- |
2, 3 |
515 |
|
AK148947.1 |
- |
2, 3 |
517 |
|
AK165683.1 |
- |
2, 3 |
487 |
|
AK197640.1 |
- |
1, 2 |
206 |
|
AK217412.1 |
- |
1, 2 |
113 |
|
AK218971.1 |
- |
1, 2 |
122 |
|
AY164700.1 |
- |
2, 3 |
550 |
|
AY427822.1 |
- |
3, 4 |
2661 |
|
AY507126.1 |
- |
2, 3 |
348 |
|
AY507127.1 |
- |
2, 3 |
2283 |
|
AY750849.1 |
- |
4, 5 |
2718 |
|
BC005478.1 |
- |
2, 3 |
459 |
|
BC014697.1 |
- |
2, 3 |
467 |
|
BC036717.1 |
- |
2, 3 |
465 |
|
BC079882.1 |
- |
4, 5 |
2647 |
6839323 |
BI729783.1 |
- |
|
527 |
|
View Full Product Details