Gene Transcripts

Gene Symbol
Lrrc8c
Entrez Gene ID
100604
Gene Name
leucine rich repeat containing 8 family, member C
Gene Aliases
AD158, AI326115, E430036I04Rik, fad158
Chromosome Location
Chr. 5: 105519386 - 105613018 on Build GRCm38
UniGene ID
Mm.319847
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133897.2 NP_598658.1 4 462
GenBank mRNA AB081508.1 - 4 462
AK089024.1 - 4 530
AK142337.1 - 4 492
AK169492.1 - 4 525
AK171313.1 - 4 544
BC026572.1 - 4 472 4988595

View Full Product Details