Gene Transcripts

Gene Symbol
SEPHS2
Entrez Gene ID
22928
Gene Name
selenophosphate synthetase 2
Gene Aliases
SPS2, SPS2b
Chromosome Location
Chr. 16: 30443625 - 30445975 on Build GRCh38
UniGene ID
Hs.118725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012248.3 NP_036380.2 1 1123
GenBank mRNA AK092003.1 - 3 875
AK093889.1 - 2 880
AK303275.1 - 2 935
BC002381.2 - 1 1007 2961275
BC016643.1 - 1 996 3923979
U43286.1 - 1 1013

View Full Product Details