Gene Transcripts

Gene Symbol
Tm4sf4
Entrez Gene ID
229302
Gene Name
transmembrane 4 superfamily member 4
Gene Aliases
Iltmp
Chromosome Location
Chr. 3: 57425410 - 57441677 on Build GRCm38
UniGene ID
Mm.26618
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145539.2 NP_663514.2 1 310
GenBank mRNA AK148484.1 - 1 278
AK165731.1 - 1 278
BB840196.1 - 1 407
BC010814.1 - 1 261 4211816

View Full Product Details